Learn More
Biomatik Corporation PGS1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09325
Encompass_Preferred
Pgs1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PGS1. The Pgs1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Cdp-Diacylglycerol--Glycerol-3-Phosphate 3-Phosphatidyltransferase, Mitochondrial Protein (110-400Aa). Applications: ELISA, WB, IHC. Target Synonyms: PGS1. Accession Number: Q32NB8. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8907