Learn More
Biomatik Corporation BBS9 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09326
Encompass_Preferred
The BBS9 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Protein PTHB1 protein (8-185AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Protein PTHB1, Bardet-Biedl syndrome 9 protein, Parathyroid hormone-responsive B1 gene protein, BBS9, PTHB1. UniProt: Q3SYG4. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8908