Learn More
Biomatik Corporation TIGIT anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09330
Encompass_Preferred
Tigit Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TIGIT. The Tigit Polyclonal Antibody has been validated for the following applications: ELISA, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human T-Cell Immunoreceptor With Ig And Itim Domains Protein (22-141Aa). Applications: ELISA, IF. Target Synonyms: TIGIT. Accession Number: Q495A1. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8912