Learn More
Biomatik Corporation HSD17B12 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09340
Encompass_Preferred
The HSD17B12 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Very-long-chain 3-oxoacyl-CoA reductase protein (40-181AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Very-long-chain 3-oxoacyl-CoA reductaseCurated (EC:1.1.1.3301 Publication, HSD17B12, 17-beta-hydroxysteroid dehydrogenase 121. UniProt: Q53GQ0. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8922