Learn More
Biomatik Corporation pat anti-Streptomyces viridochromogenes, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09345
Encompass_Preferred
Pat Polyclonal Antibody is a Streptomyces viridochromogenes Polyclonal Antibody to pat. The Pat Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Streptomyces viridochromogenes. Immunogen: Recombinant Streptomyces Viridochromogenes Phosphinothricin N-Acetyltransferase Protein (1-183Aa). Applications: ELISA, WB. Target Synonyms: pat. Accession Number: Q57146. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8927