Learn More
Biomatik Corporation FNDC3B anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09348
Encompass_Preferred
The FNDC3B Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Fibronectin type III domain-containing protein 3B protein (1-512AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Fibronectin type III domain-containing protein 3B, FNDC3B, FAD104, NS5ABP37, UNQ2421/PRO4979/PRO34274, Factor for adipocyte differentiation 104, HCV NS5A-binding protein 37. UniProt: Q53EP0. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8930