Learn More
Biomatik Corporation TM4SF20 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508931
Encompass_Preferred
Tm4Sf20 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TM4SF20. The Tm4Sf20 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Transmembrane 4 L6 Family Member 20 Protein (114-184Aa). Applications: ELISA, IHC, IF. Target Synonyms: TM4SF20. Accession Number: Q53R12. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8931 Supplier Biomatik Corporation Supplier No. CAC09349
Tm4Sf20 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TM4SF20. The Tm4Sf20 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Transmembrane 4 L6 Family Member 20 Protein (114-184Aa). Applications: ELISA, IHC, IF. Target Synonyms: TM4SF20. Accession Number: Q53R12. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.