Learn More
Biomatik Corporation GLT8D1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09354
Encompass_Preferred
The GLT8D1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Glycosyltransferase 8 domain-containing protein 1 protein (33-332AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Glycosyltransferase 8 domain-containing protein 1, GLT8D1, GALA4A, AD-017, MSTP137, UNQ572/PRO1134. UniProt: Q68CQ7. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8936