Learn More
Biomatik Corporation SCARA3 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508944
Encompass_Preferred
Scara3 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SCARA3. The Scara3 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Scavenger Receptor Class A Member 3 Protein (201-466Aa). Applications: ELISA, WB, IHC. Target Synonyms: SCARA3. Accession Number: Q6AZY7. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8944 Supplier Biomatik Corporation Supplier No. CAC09362
Scara3 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SCARA3. The Scara3 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Scavenger Receptor Class A Member 3 Protein (201-466Aa). Applications: ELISA, WB, IHC. Target Synonyms: SCARA3. Accession Number: Q6AZY7. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.