Learn More
Biomatik Corporation PVRIG anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09363
Encompass_Preferred
Pvrig Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PVRIG. The Pvrig Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Transmembrane Protein Pvrig Protein (193-326Aa). Applications: ELISA, WB, IF. Target Synonyms: PVRIG. Accession Number: Q6DKI7. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8945