Learn More
Biomatik Corporation DUSP13 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier: Biomatik Corporation CAC09376
The DUSP13 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant human Dual specificity protein phosphatase 13 isoform A protein (1-198AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-5000, IHC:1:500-1:1000. Alternative Names: DUSP13A, ranching-enzyme interacting DSP, Muscle-restricted DSP, MDSP, DUSP13, BEDP, DUSP13A, MDSP. UniProt: Q6B8I1. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.