Learn More
Biomatik Corporation CYB5R2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09377
Encompass_Preferred
The CYB5R2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human NADH-cytochrome b5 reductase 2 protein (1-237AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: NADH-cytochrome b5 reductase 2, b5R.2, CYB5R2. UniProt: Q6BCY4. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8959