Learn More
Biomatik Corporation ATG14 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09382
Encompass_Preferred
The ATG14 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant Human Beclin 1-associated autophagy-related key regulator protein (1-492AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:200-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Beclin 1-associated autophagy-related key regulator1, Autophagy-related protein 14-like protein, ATG14. UniProt: Q6ZNE5. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8964