Learn More
Biomatik Corporation SFXN4 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09386
Encompass_Preferred
Sfxn4 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SFXN4. The Sfxn4 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Sideroflexin-4 Protein (2-110Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SFXN4. Accession Number: Q6P4A7. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8968