Learn More
Biomatik Corporation LRRC31 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09389
Encompass_Preferred
Lrrc31 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LRRC31. The Lrrc31 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Leucine-Rich Repeat-Containing Protein 31 Protein (1-300Aa). Applications: ELISA, WB, IHC. Target Synonyms: LRRC31. Accession Number: Q6UY01. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8971