Learn More
Biomatik Corporation C5orf45 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508977
Encompass_Preferred
The C5orf45 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human UPF0544 protein C5orf45 protein (1-200AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: UPF0544 protein C5orf45, C5orf45. UniProt: Q6NTE8. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8977 Supplier Biomatik Corporation Supplier No. CAC09395
The C5orf45 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human UPF0544 protein C5orf45 protein (1-200AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: UPF0544 protein C5orf45, C5orf45. UniProt: Q6NTE8. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.