Learn More
Biomatik Corporation DHX32 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09399
Encompass_Preferred
The DHX32 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX32 protein (301-500AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200. Alternative Names: Putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX32, DEAD/H box 32, DEAD/H helicase-like protein 1, DHLP1, DEAH box protein 32, HuDDX32, DHX32, DDX32. UniProt: Q7L7V1. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8981