Learn More
Biomatik Corporation Defb14 anti-Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09400
Encompass_Preferred
The Defb14 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse, Rat. Immunogen: Recombinant mouse Beta-defensin 14 protein (23-67AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: BD-14, mBD-14, Defensin, beta 14, Defb14. UniProt: Q7TNV9. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8982