Learn More
Biomatik Corporation GFRAL anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09402
Encompass_Preferred
The GFRAL Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human GDNF family receptor alpha-like protein (19-351AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-5000, IHC:1:20-1:200. Alternative Names: GDNF family receptor alpha-like, GFRAL, C6orf144, UNQ9356/PRO34128. UniProt: Q6UXV0. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8984