Learn More
Biomatik Corporation CNIH2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09407
Encompass_Preferred
The CNIH2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Protein cornichon homolog 2 protein (94-138AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Protein cornichon homolog 2, CNIH-2, Cornichon family AMPA receptor auxiliary protein 2, Cornichon-like protein, CNIH2, CNIL. UniProt: Q6PI25. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8989