Learn More
Biomatik Corporation CFAP65 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09411
Encompass_Preferred
The CFAP65 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Coiled-coil domain-containing protein 108 protein (2-164AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Coiled-coil domain-containing protein 108, CCDC108. UniProt: Q6ZU64. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8993