Learn More
Biomatik Corporation inlA anti-Listeria monocytogenes serotype 4b, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501508994
Encompass_Preferred
Inla Polyclonal Antibody is a Listeria monocytogenes serotype 4b Polyclonal Antibody to inlA. The Inla Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Listeria monocytogenes serotype 4b. Immunogen: Recombinant Listeria Monocytogenes Serotype 4B Internalin-A Protein (562-770Aa). Applications: ELISA, WB. Target Synonyms: inlA. Accession Number: Q723K6. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8994 Supplier Biomatik Corporation Supplier No. CAC09412
Inla Polyclonal Antibody is a Listeria monocytogenes serotype 4b Polyclonal Antibody to inlA. The Inla Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Listeria monocytogenes serotype 4b. Immunogen: Recombinant Listeria Monocytogenes Serotype 4B Internalin-A Protein (562-770Aa). Applications: ELISA, WB. Target Synonyms: inlA. Accession Number: Q723K6. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.