Learn More
Biomatik Corporation SLC25A25 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09414
Encompass_Preferred
Slc25A25 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SLC25A25. The Slc25A25 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Calcium-Binding Mitochondrial Carrier Protein Scamc-2 Protein (1-189Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SLC25A25. Accession Number: Q6KCM7. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8996