Learn More
Biomatik Corporation C16orf87 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09423
Encompass_Preferred
The C16orf87 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human UPF0547 protein C16orf87 protein (1-154AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: UPF0547 protein C16orf87, C16orf87. UniProt: Q6PH81. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9005