Learn More
Biomatik Corporation PKD1L3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09426
Encompass_Preferred
Pkd1L3 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PKD1L3. The Pkd1L3 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Polycystic Kidney Disease Protein 1-Like 3 Protein (22-194Aa). Applications: ELISA, IHC, IF. Target Synonyms: PKD1L3. Accession Number: Q7Z443. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9008