Learn More
Biomatik Corporation PLCZ1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509011
Encompass_Preferred
Plcz1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PLCZ1. The Plcz1 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human 1-Phosphatidylinositol 4,5-Bisphosphate Phosphodiesterase Zeta-1 Protein (1-415Aa). Applications: ELISA, WB. Target Synonyms: PLCZ1. Accession Number: Q86YW0. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9011 Supplier Biomatik Corporation Supplier No. CAC09429
Plcz1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PLCZ1. The Plcz1 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human 1-Phosphatidylinositol 4,5-Bisphosphate Phosphodiesterase Zeta-1 Protein (1-415Aa). Applications: ELISA, WB. Target Synonyms: PLCZ1. Accession Number: Q86YW0. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.