Learn More
Biomatik Corporation TRPM8 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09430
Encompass_Preferred
Trpm8 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TRPM8. The Trpm8 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Transient Receptor Potential Cation Channel Subfamily M Member 8 Protein (1-192Aa). Applications: ELISA, IHC, IF. Target Synonyms: TRPM8. Accession Number: Q7Z2W7. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9012