Learn More
Biomatik Corporation LYSMD3 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509014
Encompass_Preferred
Lysmd3 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LYSMD3. The Lysmd3 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Lysm And Putative Peptidoglycan-Binding Domain-Containing Protein 3 Protein (1-127Aa). Applications: ELISA, WB. Target Synonyms: LYSMD3. Accession Number: Q7Z3D4. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9014 Supplier Biomatik Corporation Supplier No. CAC09432
Lysmd3 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LYSMD3. The Lysmd3 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Lysm And Putative Peptidoglycan-Binding Domain-Containing Protein 3 Protein (1-127Aa). Applications: ELISA, WB. Target Synonyms: LYSMD3. Accession Number: Q7Z3D4. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.