Learn More
Biomatik Corporation ISCA2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09434
Encompass_Preferred
Isca2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to ISCA2. The Isca2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Iron-Sulfur Cluster Assembly 2 Homolog, Mitochondrial Protein (9-154Aa). Applications: ELISA, WB, IHC. Target Synonyms: ISCA2. Accession Number: Q86U28. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9016