Learn More
Biomatik Corporation GALNT7 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09438
Encompass_Preferred
The GALNT7 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human N-acetylgalactosaminyltransferase 7 protein (201-500AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: N-acetylgalactosaminyltransferase 7, Polypeptide GalNAc transferase 7, GalNAc-T7, pp-GaNTase 7, Protein-UDP acetylgalactosaminyltransferase 7, polypeptide N-acetylgalactosaminyltransferase 7, GALNT7. UniProt: Q86SF2. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9020