Learn More
Biomatik Corporation UNC93A anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509021
Encompass_Preferred
Unc93A Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to UNC93A. The Unc93A Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Protein Unc-93 Homolog A Protein (374-415Aa). Applications: ELISA, WB, IHC. Target Synonyms: UNC93A. Accession Number: Q86WB7. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9021 Supplier Biomatik Corporation Supplier No. CAC09439
Unc93A Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to UNC93A. The Unc93A Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Protein Unc-93 Homolog A Protein (374-415Aa). Applications: ELISA, WB, IHC. Target Synonyms: UNC93A. Accession Number: Q86WB7. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.