Learn More
Biomatik Corporation ERO1B anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09445
Encompass_Preferred
The ERO1B Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human ERO1-like protein beta protein (101-400AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: ERO1-like protein beta, ERO1-L-beta, Endoplasmic reticulum oxidoreductase beta, ERO1B. UniProt: Q86YB8. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9027