Learn More
Biomatik Corporation ANKRD23 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09448
Encompass_Preferred
The ANKRD23 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Ankyrin repeat domain-containing protein 23 protein (1-200AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200. Alternative Names: Ankyrin repeat domain-containing protein 23, Diabetes-related ankyrin repeat protein, Muscle ankyrin repeat protein 3, ANKRD23, DARP. UniProt: Q86SG2. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9030