Learn More
Biomatik Corporation TPH2 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509034
Encompass_Preferred
Tph2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TPH2. The Tph2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Tryptophan 5-Hydroxylase 2 Protein (44-225Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: TPH2. Accession Number: Q8IWU9. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9034 Supplier Biomatik Corporation Supplier No. CAC09452
Tph2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TPH2. The Tph2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Tryptophan 5-Hydroxylase 2 Protein (44-225Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: TPH2. Accession Number: Q8IWU9. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.