Learn More
Biomatik Corporation EGFL6 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09459
Encompass_Preferred
The EGFL6 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Epidermal growth factor-like protein 6 protein (301-554AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Epidermal growth factor-like protein 6, EGF-like protein 6, EGFL6, MAEG, PP648, UNQ281/PRO320, MAM and EGF domains-containing gene protein. UniProt: Q8IUX8. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9041