Learn More
Biomatik Corporation ZBTB8OS anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09471
Encompass_Preferred
Zbtb8Os Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to ZBTB8OS. The Zbtb8Os Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Protein Archease Protein (2-136Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: ZBTB8OS. Accession Number: Q8IWT0. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9053