Learn More
Biomatik Corporation KIAA0319L anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09473
Encompass_Preferred
Kiaa0319L Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to KIAA0319L. The Kiaa0319L Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Dyslexia-Associated Protein Kiaa0319-Like Protein (479-718Aa). Applications: ELISA, IHC, IF. Target Synonyms: KIAA0319L. Accession Number: Q8IZA0. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9055