Learn More
Biomatik Corporation SLC38A6 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09478
Encompass_Preferred
Slc38A6 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SLC38A6. The Slc38A6 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Probable Sodium-Coupled Neutral Amino Acid Transporter 6 Protein (132-169Aa). Applications: ELISA, WB, IF. Target Synonyms: SLC38A6. Accession Number: Q8IZM9. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9060