Learn More
Biomatik Corporation FAHD2A anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09493
Encompass_Preferred
The FAHD2A Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Fumarylacetoacetate hydrolase domain-containing protein 2A protein (4-314AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Fumarylacetoacetate hydrolase domain-containing protein 2A, FAHD2A, CGI-105. UniProt: Q96GK7. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9075