Learn More
Biomatik Corporation C4orf33 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09502
Encompass_Preferred
The C4orf33 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human UPF0462 protein C4orf33 protein (1-199AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: UPF0462 protein C4orf33, C4orf33. UniProt: Q8N1A6. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9084