Learn More
Biomatik Corporation LIX1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509089
Encompass_Preferred
Lix1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LIX1. The Lix1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Protein Limb Expression 1 Homolog Protein (1-282Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: LIX1. Accession Number: Q8N485. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9089 Supplier Biomatik Corporation Supplier No. CAC09507
Lix1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LIX1. The Lix1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Protein Limb Expression 1 Homolog Protein (1-282Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: LIX1. Accession Number: Q8N485. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.