Learn More
Biomatik Corporation TRIML1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09511
Encompass_Preferred
Triml1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TRIML1. The Triml1 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Probable E3 Ubiquitin-Protein Ligase Triml1 Protein (277-468Aa). Applications: ELISA, WB. Target Synonyms: TRIML1. Accession Number: Q8N9V2. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9093