Learn More
Biomatik Corporation UXS1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09513
Encompass_Preferred
Uxs1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to UXS1. The Uxs1 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Udp-Glucuronic Acid Decarboxylase 1 Protein (201-420Aa). Applications: ELISA, WB. Target Synonyms: UXS1. Accession Number: Q8NBZ7. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9095