Learn More
Biomatik Corporation GSDMB anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509098
Encompass_Preferred
The GSDMB Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Gasdermin-B protein (63-310AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Gasdermin-B, GSDMB, GSDML, PP4052, PRO2521, Gasdermin-like protein. UniProt: Q8TAX9. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9098 Supplier Biomatik Corporation Supplier No. CAC09516
The GSDMB Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Gasdermin-B protein (63-310AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Gasdermin-B, GSDMB, GSDML, PP4052, PRO2521, Gasdermin-like protein. UniProt: Q8TAX9. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.