Learn More
Biomatik Corporation DIS3L anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509100
Encompass_Preferred
The DIS3L Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human DIS3-like exonuclease 1 protein (1-300AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: DIS3-like exonuclease 1 (EC:3.1.13.-), DIS3L, DIS3L1, KIAA1955. UniProt: Q8TF46. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9100 Supplier Biomatik Corporation Supplier No. CAC09518
The DIS3L Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human DIS3-like exonuclease 1 protein (1-300AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: DIS3-like exonuclease 1 (EC:3.1.13.-), DIS3L, DIS3L1, KIAA1955. UniProt: Q8TF46. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.