Learn More
Biomatik Corporation RGMA anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509107
Encompass_Preferred
Rgma Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RGMA. The Rgma Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Repulsive Guidance Molecule A Protein (153-408Aa). Applications: ELISA, WB, IHC. Target Synonyms: RGMA. Accession Number: Q96B86. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9107 Supplier Biomatik Corporation Supplier No. CAC09525
Rgma Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RGMA. The Rgma Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Repulsive Guidance Molecule A Protein (153-408Aa). Applications: ELISA, WB, IHC. Target Synonyms: RGMA. Accession Number: Q96B86. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.