Learn More
Biomatik Corporation MRPL48 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509111
Encompass_Preferred
Mrpl48 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MRPL48. The Mrpl48 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human 39S Ribosomal Protein L48, Mitochondrial Protein (29-212Aa). Applications: ELISA, WB, IHC. Target Synonyms: MRPL48. Accession Number: Q96GC5. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9111 Supplier Biomatik Corporation Supplier No. CAC09529
Mrpl48 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MRPL48. The Mrpl48 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human 39S Ribosomal Protein L48, Mitochondrial Protein (29-212Aa). Applications: ELISA, WB, IHC. Target Synonyms: MRPL48. Accession Number: Q96GC5. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.