Learn More
Biomatik Corporation PRR11 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509113
Encompass_Preferred
Prr11 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PRR11. The Prr11 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Proline-Rich Protein 11 Protein (1-360Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PRR11. Accession Number: Q96HE9. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9113 Supplier Biomatik Corporation Supplier No. CAC09531
Prr11 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PRR11. The Prr11 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Proline-Rich Protein 11 Protein (1-360Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PRR11. Accession Number: Q96HE9. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.