Learn More
Biomatik Corporation HDDC3 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09533
Encompass_Preferred
The HDDC3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Guanosine-3',5'-bis (diphosphate) 3'-pyrophosphohydrolase MESH1 protein (2-140AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: HD domain-containing protein 3, Metazoan SpoT homolog 1, MESH1, Penta-phosphate guanosine-3'-pyrophosphohydrolase, (ppGpp)ase, MESH1, HDDC3. UniProt: Q8N4P3. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9115