Learn More
Biomatik Corporation ATL2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509119
Encompass_Preferred
The ATL2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Atlastin-2 protein (1-300AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Atlastin-2, ADP-ribosylation factor-like protein 6-interacting protein 2, ARL-6-interacting protein 2, Aip-2, ATL2, ARL6IP2. UniProt: Q8NHH9. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9119 Supplier Biomatik Corporation Supplier No. CAC09537
The ATL2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Atlastin-2 protein (1-300AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Atlastin-2, ADP-ribosylation factor-like protein 6-interacting protein 2, ARL-6-interacting protein 2, Aip-2, ATL2, ARL6IP2. UniProt: Q8NHH9. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.